Gene Transcripts
- Gene Symbol
- TMCO5A
- Entrez Gene ID
- 145942
- Gene Name
- transmembrane and coiled-coil domains 5A
- Gene Aliases
- TMCO5
- Chromosome Location
- Chr. 15: 37934607 - 37997541 on Build GRCh38
- UniGene ID
- Hs.179646
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152453.3 |
NP_689666.2
|
3, 4 |
338 |
|
XM_005254168.3 |
XP_005254225.1
|
3, 4 |
319 |
|
XM_011521262.2 |
XP_011519564.1
|
3, 4 |
319 |
|
XM_011521263.1 |
XP_011519565.1
|
3, 4 |
319 |
|
XM_011521264.1 |
XP_011519566.1
|
3, 4 |
319 |
|
GenBank mRNA |
AK093126.1 |
- |
3, 4 |
319 |
|
AK310517.1 |
- |
3, 4 |
324 |
|
BC029221.2 |
- |
2, 3 |
251 |
5167874 |
BI561792.1 |
- |
3, 4 |
342 |
|
BI826726.1 |
- |
3, 4 |
302 |
|
View Full Product Details