Gene Transcripts
- Gene Symbol
- CSNK1G2-AS1
- Entrez Gene ID
- 255193
- Gene Name
- CSNK1G2 antisense RNA 1
- Gene Aliases
- C19orf34
- Chromosome Location
- Chr. 19: 1952527 - 1954549 on Build GRCh38
- UniGene ID
- Hs.374717
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033400.1 |
|
1, 2 |
703 |
|
GenBank mRNA |
BC029585.1 |
- |
2, 3 |
647 |
|
View Full Product Details