Gene Transcripts

Gene Symbol
BEST4
Entrez Gene ID
266675
Gene Name
bestrophin 4
Gene Aliases
VMD2L2
Chromosome Location
Chr. 1: 44781836 - 44792828 on Build GRCh38
UniGene ID
Hs.302513
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153274.2 NP_695006.1 2 233
XM_017001020.1 XP_016856509.1 2 1481
XM_017001021.1 XP_016856510.1 4 460
XM_017001022.1 XP_016856511.1 4 1584
XM_017001023.1 XP_016856512.1 4 1584
GenBank mRNA AF440757.1 - 2 184
AY515707.1 - 2 184
BC101823.1 - 2 233 8069329

View Full Product Details