Gene Transcripts

Gene Symbol
ALDH5A1
Entrez Gene ID
7915
Gene Name
aldehyde dehydrogenase 5 family member A1
Gene Aliases
SSADH, SSDH
Chromosome Location
Chr. 6: 24494969 - 24537207 on Build GRCh38
UniGene ID
Hs.371723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080.3 NP_001071.1 2 395
NM_170740.1 NP_733936.1 2 395
GenBank mRNA AJ427353.1 - 2 163
AJ427354.2 - 2 163
AJ427355.2 - 2 367
AK290800.1 - 2 513
AK294699.1 - 3 172
AK314357.1 - 2 395
AK315380.1 - 2 490
BC034321.1 - 2 395 4829533
KJ534766.1 - 2 367
KJ534767.1 - 2 367
KJ534768.1 - 2 367
KJ534769.1 - 2 367
Y11192.1 - 2 367

View Full Product Details