Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145128.2 | NP_001138600.2 | 36 | 5038 | |
XM_006715376.3 | XP_006715439.2 | 37 | 5903 | ||
XM_011535550.2 | XP_011533852.1 | 37 | 5918 | ||
XM_011535552.2 | XP_011533854.1 | 36 | 5094 | ||
XM_011535553.2 | XP_011533855.1 | 36 | 5701 | ||
XM_011535554.2 | XP_011533856.1 | 35 | 5597 | ||
XM_011535558.1 | XP_011533860.1 | 18 | 2939 | ||
XM_017010385.1 | XP_016865874.1 | 36 | 5086 | ||
XM_017010387.1 | XP_016865876.1 | 31 | 5549 | ||
XM_017010389.1 | XP_016865878.1 | 22 | 3801 | ||
GenBank mRNA | AK098657.1 | - | 2 | 276 | |
BC101714.1 | - | 2 | 204 | 8069220 | |
BC111948.1 | - | 2 | 204 | 8327416 | |
BC143533.1 | - | 2 | 162 | 9052042 |