Gene Transcripts

Gene Symbol
LMNTD2
Entrez Gene ID
256329
Gene Name
lamin tail domain containing 2
Gene Aliases
C11orf35
Chromosome Location
Chr. 11: 554850 - 560780 on Build GRCh38
UniGene ID
Hs.669395
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173573.2 NP_775844.2 8, 9 1036
XM_011519965.2 XP_011518267.2 8, 9 1211
XM_011519967.2 XP_011518269.2 8, 9 1211
XM_017017476.1 XP_016872965.1 8, 9 1211
XM_017017477.1 XP_016872966.1 8, 9 1196
XM_017017478.1 XP_016872967.1 8, 9 1052
XM_017017479.1 XP_016872968.1 8, 9 1211
GenBank mRNA BC039077.1 - 8, 9 1036

View Full Product Details