Gene Transcripts

Gene Symbol
SEC14L4
Entrez Gene ID
284904
Gene Name
SEC14 like lipid binding 4
Gene Aliases
TAP3
Chromosome Location
Chr. 22: 30488150 - 30505711 on Build GRCh38
UniGene ID
Hs.517541
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161368.1 NP_001154840.1 4, 5 342
NM_174977.3 NP_777637.1 4, 5 342
XM_006724235.2 XP_006724298.1 4, 5 358
XM_011530164.2 XP_011528466.1 4, 5 522
XM_017028787.1 XP_016884276.1 3, 4 471
XM_017028788.1 XP_016884277.1 3, 4 472
XM_017028789.1 XP_016884278.1 3, 4 144
XM_017028790.1 XP_016884279.1 3, 4 479
XM_017028791.1 XP_016884280.1 3, 4 171
GenBank mRNA AK092002.1 - 5, 6 391
AK093418.1 - 4, 5 359
AK093448.1 - 4, 5 326
AY158085.1 - 4, 5 242
BC136358.1 - 4, 5 314 9020345
BC136359.1 - 4, 5 314 9020346
BC139912.1 - 4, 5 281 40147686
CR456346.1 - 2, 3 101

View Full Product Details