Gene Transcripts

Gene Symbol
SLC5A2
Entrez Gene ID
6524
Gene Name
solute carrier family 5 member 2
Gene Aliases
SGLT2
Chromosome Location
Chr. 16: 31482535 - 31490770 on Build GRCh38
UniGene ID
Hs.709195
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003041.3 NP_003032.1 14 2243
NR_130783.1 12 1923
GenBank mRNA AK054830.1 - 3 2438
M95549.1 - 14 2244

View Full Product Details