Gene Transcripts
- Gene Symbol
- PYY2
- Entrez Gene ID
- 23615
- Gene Name
- peptide YY, 2 (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 17: 28226563 - 28228060 on Build GRCh38
- UniGene ID
- Hs.157195
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF222904.1 |
- |
2 |
816 |
|
View Full Product Details