Gene Transcripts

Gene Symbol
ARMC5
Entrez Gene ID
79798
Gene Name
armadillo repeat containing 5
Gene Aliases
AIMAH2
Chromosome Location
Chr. 16: 31458273 - 31467167 on Build GRCh38
UniGene ID
Hs.732945
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105247.1 NP_001098717.1 2 1026
NM_001288767.1 NP_001275696.1 4 906
NM_001301820.1 NP_001288749.1 3 1041
NM_024742.2 NP_079018.1 2 1026
XM_006721091.2 XP_006721154.1 3 1159
GenBank mRNA AK023125.1 - 2 77
AK024430.1 - 2 519
AK294897.1 - 3 948
AK295790.1 - 4 906
AK297115.1 - 2 61
AY217348.1 - 2 1026
BC014945.1 - 3 431 4860377

View Full Product Details