Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024852.3 | NP_079128.2 | 2 | 503 | |
NM_177422.2 | NP_803171.1 | 2 | 503 | ||
XM_005270575.3 | XP_005270632.1 | 3 | 5957 | ||
XM_011540879.2 | XP_011539181.1 | 2 | 1385 | ||
XM_017000523.1 | XP_016856012.1 | 3 | 5973 | ||
GenBank mRNA | AK022827.1 | - | 2 | 258 | |
AK027796.1 | - | 3 | 437 | ||
AK292870.1 | - | 2 | 469 | ||
BC025769.1 | - | 2 | 327 | ||
BC066888.1 | - | 2 | 503 | ||
BF979526.1 | - | 2 | 508 |