Gene Transcripts

Gene Symbol
MKNK1
Entrez Gene ID
8569
Gene Name
MAP kinase interacting serine/threonine kinase 1
Gene Aliases
MNK1
Chromosome Location
Chr. 1: 46557407 - 46607140 on Build GRCh38
UniGene ID
Hs.371594
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135553.2 NP_001129025.1 3, 4 376
NM_003684.5 NP_003675.2 3, 4 376
NM_198973.3 NP_945324.1 3, 4 334
NR_024174.2 3, 4 376
NR_024176.2 4, 5 477
XM_006711000.1 XP_006711063.1 3, 4 329
XM_006711001.2 XP_006711064.1 4, 5 579
XM_006711002.2 XP_006711065.1 3, 4 5804
XM_006711003.1 XP_006711066.1 4, 5 390
XM_006711013.1 XP_006711076.1 3, 4 328
XM_006711014.1 XP_006711077.1 3, 4 319
XM_006711015.1 XP_006711078.1 4, 5 390
XM_017002655.1 XP_016858144.1 4, 5 6150
XM_017002656.1 XP_016858145.1 4, 5 5949
XM_017002659.1 XP_016858148.1 3, 4 348
XM_017002660.1 XP_016858149.1 3, 4 5804
XM_017002661.1 XP_016858150.1 4, 5 569
XM_017002662.1 XP_016858151.1 4, 5 390
XM_017002663.1 XP_016858152.1 4, 5 6150
XM_017002664.1 XP_016858153.1 3, 4 5804
XM_017002666.1 XP_016858155.1 4, 5 3279
XM_017002667.1 XP_016858156.1 2, 3 855
GenBank mRNA AB000409.1 - 3, 4 319
AB451300.1 - 2, 3 132
AB451432.1 - 2, 3 132
AK096423.1 - 4, 5 421
AK290456.1 - 3, 4 296
AK298839.1 - 3, 4 350
AK304005.1 - 3, 4 399
AY355461.1 - 2, 3 132
BC002755.2 - 3, 4 296 3629765
BG722049.1 - 3, 4 339
BX538193.1 - 2, 3 200
DA435648.1 - 3, 4 321
DA502363.1 - 3, 4 349
DC366212.1 - 3, 4 350
DC376088.1 - 3, 4 306

View Full Product Details