Gene Transcripts
- Gene Symbol
- C1QTNF5
- Entrez Gene ID
- 114902
- Gene Name
- C1q and tumor necrosis factor related protein 5
- Gene Aliases
- CTRP5
- Chromosome Location
- Chr. 11: 119338934 - 119346673 on Build GRCh38
- UniGene ID
- Hs.632102
- Species
- Homo sapiens
- Gene Symbol
- MFRP
- Entrez Gene ID
- 83552
- Gene Name
- membrane frizzled-related protein
- Gene Aliases
- MCOP5, NNO2, RD6
- Chromosome Location
- Chr. 11: 119338934 - 119346673 on Build GRCh38
- UniGene ID
- Hs.632102
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031433.3 |
NP_113621.1
|
15 |
3299 |
|
GenBank mRNA |
AJ862823.1 |
- |
15 |
3281 |
|
View Full Product Details