Gene Transcripts

Gene Symbol
ASMTL-AS1
Entrez Gene ID
80161
Gene Name
ASMTL antisense RNA 1
Gene Aliases
ASMTL-AS, ASMTLAS, CXYorf2, NCRNA00105
Chromosome Location
Chr. Y: 1400531 - 1415421 on Build GRCh38
UniGene ID
Hs.521856
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026710.1 4 1061
NR_026711.1 4 1061
GenBank mRNA AF193043.2 - 3 424
AK023392.1 - 4 494
BC093994.2 - 3 404
BC143527.1 - 3 438

View Full Product Details