Gene Transcripts

Gene Symbol
Alox5
Entrez Gene ID
25290
Gene Name
arachidonate 5-lipoxygenase
Gene Aliases
LOX5A
Chromosome Location
Chr. 4: 148398004 - 148446308 on Build Rnor_6.0
UniGene ID
Rn.9662
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012822.1 NP_036954.1 7 934
XM_006237140.2 XP_006237202.1 7 971
GenBank mRNA J03960.1 - 7 934

View Full Product Details