Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001316361.2 | NP_001303290.1 | 4 | 607 | |
NM_001323347.1 | NP_001310276.1 | 4 | 320 | ||
NM_003657.3 | NP_003648.2 | 4 | 607 | ||
XM_005260587.2 | XP_005260644.1 | 4 | 387 | ||
XM_005260589.2 | XP_005260646.1 | 4 | 387 | ||
XM_005260590.2 | XP_005260647.1 | 4 | 387 | ||
XM_005260591.2 | XP_005260648.1 | 4 | 387 | ||
XM_005260593.2 | XP_005260650.1 | 4 | 387 | ||
XM_005260594.2 | XP_005260651.1 | 4 | 387 | ||
XM_005260595.2 | XP_005260652.1 | 4 | 387 | ||
XM_011529090.2 | XP_011527392.1 | 4 | 315 | ||
XM_011529091.2 | XP_011527393.1 | 4 | 387 | ||
XM_017028110.1 | XP_016883599.1 | 4 | 387 | ||
XM_017028111.1 | XP_016883600.1 | 4 | 387 | ||
XM_017028112.1 | XP_016883601.1 | 4 | 387 | ||
GenBank mRNA | AF041260.1 | - | 4 | 386 | |
AK225656.1 | - | 4 | 376 | ||
AK304213.1 | - | 2 | 186 | ||
AK308384.1 | - | 4 | 376 | ||
AK315220.1 | - | 4 | 607 | ||
BC024044.1 | - | 4 | 387 | 4645428 | |
BC126346.1 | - | 4 | 318 | 8992062 | |
BC126348.1 | - | 4 | 318 | 8992064 | |
DC358921.1 | - | 4 | 320 |