Gene Transcripts

Gene Symbol
Mndal
Entrez Gene ID
100040462
Gene Name
myeloid nuclear differentiation antigen like
Gene Aliases
Gm2785
Chromosome Location
Chr. 1: 173857220 - 173880260 on Build GRCm38
UniGene ID
Mm.463437
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170853.1 NP_001164324.1 104
GenBank mRNA AK136391.1 - 105

Gene Symbol
Ifi203
Entrez Gene ID
15950
Gene Name
interferon activated gene 203
Gene Aliases
Chromosome Location
Chr. 1: 173920400 - 173942847 on Build GRCm38
UniGene ID
Mm.463437
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF022371.1 - 165
AK169754.1 - 2, 3 99
BC008167.1 - 207 3257492

View Full Product Details