Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033202.3 | NP_001028374.1 | 13 | 2041 | |
GenBank mRNA | AK078164.1 | - | 9 | 1393 | |
AK144578.1 | - | 13 | 2042 | ||
AK162153.1 | - | 1090 | |||
AK215668.1 | - | 1 | 218 | ||
BC038606.1 | - | 13 | 2063 |