Gene Transcripts

Gene Symbol
Lama5
Entrez Gene ID
16776
Gene Name
laminin, alpha 5
Gene Aliases
AA408760, AA408762, AI853660, [a]5, laminin-511, mKIAA0533
Chromosome Location
Chr. 2: 180176373 - 180225905 on Build GRCm38
UniGene ID
Mm.4339
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081171.2 NP_001074640.1 73 10231
GenBank mRNA AK129157.1 - 34 5005
AK158204.1 - 6 858
BC020313.2 - 10 1403
BC043478.1 - 3 377
BC062207.1 - 11 1664
CN702366.1 - 2 220
U37501.1 - 73 9925

View Full Product Details