Gene Transcripts
- Gene Symbol
- Btbd2
- Entrez Gene ID
- 208198
- Gene Name
- BTB (POZ) domain containing 2
- Gene Aliases
- 2610037C03Rik, 4930512K17Rik
- Chromosome Location
- Chr. 10: 80642617 - 80657118 on Build GRCm38
- UniGene ID
- Mm.60720
- Species
- Mus musculus
- Gene Symbol
- Zkscan17
- Entrez Gene ID
- 268417
- Gene Name
- zinc finger with KRAB and SCAN domains 17
- Gene Aliases
- Nizp1, Zfp496, Znf496
- Chromosome Location
- Chr. 11: 59485520 - 59506640 on Build GRCm38
- UniGene ID
- Mm.239793
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK182740.1 |
- |
|
17 |
|
View Full Product Details