Gene Transcripts

Gene Symbol
Btbd2
Entrez Gene ID
208198
Gene Name
BTB (POZ) domain containing 2
Gene Aliases
2610037C03Rik, 4930512K17Rik
Chromosome Location
Chr. 10: 80642617 - 80657118 on Build GRCm38
UniGene ID
Mm.60720
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145361.2 NP_663336.2 7 1782
GenBank mRNA AK046604.1 - 5 630
AK194965.1 - 2 87
BC016566.1 - 3 328
BC055704.1 - 6 849
BC069864.1 - 3 301
BC083320.1 - 6 755
BC115682.1 - 6 842 40104217
BC115683.1 - 6 842 40104219

View Full Product Details