Gene Transcripts

Gene Symbol
Lactbl1
Entrez Gene ID
242707
Gene Name
lactamase, beta-like 1
Gene Aliases
Chromosome Location
Chr. 4: 136622621 - 136638110 on Build GRCm38
UniGene ID
Mm.448979
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243262.1 NP_001230191.1 1 44
GenBank mRNA BC028950.1 - 44
BC029684.1 - 1 73

View Full Product Details