Gene Transcripts

Gene Symbol
Ankdd1b
Entrez Gene ID
271144
Gene Name
ankyrin repeat and death domain containing 1B
Gene Aliases
9330128J19Rik
Chromosome Location
Chr. 13: 96415030 - 96471354 on Build GRCm38
UniGene ID
Mm.470057
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042714.1 NP_001036179.1 11 1170
GenBank mRNA AK033386.1 - 5 410
AK033960.1 - 4 364
BC112417.1 - 11 1167 40061167

View Full Product Details