Gene Transcripts

Gene Symbol
B3gnt6
Entrez Gene ID
272411
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6 (core 3 synthase)
Gene Aliases
BC039789, Core-3, Core3
Chromosome Location
Chr. 7: 98192415 - 98199475 on Build GRCm38
UniGene ID
Mm.98333
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081167.1 NP_001074636.1 2 1490
GenBank mRNA AK140429.1 - 1 1064
BC039789.1 - 2 1506

View Full Product Details