Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163175.1 | NP_001156647.1 | 7 | 2185 | |
GenBank mRNA | AK158915.1 | - | 7 | 2187 | |
BC059879.1 | - | 1 | 2192 | ||
BC080282.1 | - | 5 | 1978 | ||
BC108424.1 | - | 1 | 2075 |