Gene Transcripts

Gene Symbol
Aes
Entrez Gene ID
29466
Gene Name
amino-terminal enhancer of split
Gene Aliases
Grg, Grg-5
Chromosome Location
Chr. 7: 11012843 - 11018156 on Build Rnor_6.0
UniGene ID
Rn.11495
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019220.1 NP_062093.1 2, 3 267
GenBank mRNA BC101855.1 - 232
L14462.1 - 2, 3 267

View Full Product Details