Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081364.3 | NP_001074833.3 | 7 | 1507 | |
NM_001128084.2 | NP_001121556.2 | 8 | 1537 | ||
GenBank mRNA | BC076629.1 | - | 7 | 1507 | |
BC150741.1 | - | 8 | 1466 | 9087652 |