Gene Transcripts

Gene Symbol
Usp31
Entrez Gene ID
76179
Gene Name
ubiquitin specific peptidase 31
Gene Aliases
6330567E21Rik, mKIAA1203
Chromosome Location
Chr. 7: 121642021 - 121707344 on Build GRCm38
UniGene ID
Mm.122538
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033173.1 NP_001028345.1 16 5915
GenBank mRNA AK018244.1 - 1 600
AK122466.1 - 1 2735

View Full Product Details