Gene Transcripts

Gene Symbol
Armc6
Entrez Gene ID
76813
Gene Name
armadillo repeat containing 6
Gene Aliases
2410153K17Rik, AW554412
Chromosome Location
Chr. 8: 70220172 - 70234422 on Build GRCm38
UniGene ID
Mm.383251
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133972.2 NP_598733.2 7 1439
GenBank mRNA AK049351.1 - 7 1155
AK080609.1 - 7 1144
AK081550.1 - 7 1484
BC005754.1 - 4 648
BC030077.1 - 4 468
BC043070.1 - 7 1439 6403587
BC048396.1 - 7 1401
BC055779.1 - 7 1439 6403587

View Full Product Details