Gene Transcripts
- Gene Symbol
- Prl8a3
- Entrez Gene ID
- 100361884
- Gene Name
- prolactin family 8, subfamily a, member 3
- Gene Aliases
- Plp-Cv, Plpc, Plpcv, Prlpc, Prlpc1
- Chromosome Location
- Chr. 17: 39455865 - 39463715 on Build Rnor_6.0
- UniGene ID
- Rn.202972
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020079.1 |
NP_064464.1
|
|
189 |
|
GenBank mRNA |
U93351.1 |
- |
|
189 |
|
View Full Product Details