Gene Transcripts

Gene Symbol
Lypd3
Entrez Gene ID
60378
Gene Name
Ly6/Plaur domain containing 3
Gene Aliases
C4.4a
Chromosome Location
Chr. 1: 81499856 - 81504306 on Build Rnor_6.0
UniGene ID
Rn.38434
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021759.1 NP_068527.1 4 604
GenBank mRNA AJ001043.1 - 4 604

View Full Product Details