Gene Transcripts

Gene Symbol
Arf2
Entrez Gene ID
79119
Gene Name
ADP-ribosylation factor 2
Gene Aliases
Chromosome Location
Chr. 10: 92018562 - 92040370 on Build Rnor_6.0
UniGene ID
Rn.62175
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024150.2 NP_077064.1 4, 5 465
XM_006247543.2 XP_006247605.1 4, 5 563
GenBank mRNA BC098915.1 - 4, 5 465
L12381.1 - 3, 4 512

View Full Product Details