Gene Transcripts

Gene Symbol
Asip
Entrez Gene ID
24152
Gene Name
agouti signaling protein
Gene Aliases
Chromosome Location
Chr. 3: 150492010 - 150579870 on Build Rnor_6.0
UniGene ID
Rn.205372
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052979.1 NP_443211.1 2 201
GenBank mRNA AB045587.1 - 2 201

View Full Product Details