Gene Transcripts

Gene Symbol
Syt8
Entrez Gene ID
60566
Gene Name
synaptotagmin VIII
Gene Aliases
Chromosome Location
Chr. 1: 215605408 - 215607602 on Build Rnor_6.0
UniGene ID
Rn.46519
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053325.1 NP_445777.1 2 229
GenBank mRNA AF375462.1 - 2 229
AF375467.1 - 2 229

View Full Product Details