Gene Transcripts

Gene Symbol
ABCD4
Entrez Gene ID
5826
Gene Name
ATP binding cassette subfamily D member 4
Gene Aliases
ABC41, EST352188, MAHCJ, P70R, P79R, PMP69, PXMP1L
Chromosome Location
Chr. 14: 74285277 - 74303064 on Build GRCh38
UniGene ID
Hs.94395
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005050.3 NP_005041.1 12 1375
NR_003256.2 11 1247
XM_005267939.3 XP_005267996.1 12 1303
XM_005267940.3 XP_005267997.1 11 1182
XM_005267941.4 XP_005267998.1 11 1137
XM_005267942.4 XP_005267999.1 10 1024
XM_005267946.3 XP_005268003.1 10 1019
XM_005267949.3 XP_005268006.1 11 1233
XM_006720223.2 XP_006720286.1 11 1187
XM_011537041.2 XP_011535343.1 10 1045
XM_017021531.1 XP_016877020.1 12 1308
XM_017021532.1 XP_016877021.1 11 1182
XM_017021533.1 XP_016877022.1 11 1182
XM_017021534.1 XP_016877023.1 10 989
XM_017021535.1 XP_016877024.1 11 1213
XM_017021536.1 XP_016877025.1 10 1033
XM_017021537.1 XP_016877026.1 9 916
XM_017021538.1 XP_016877027.1 10 1034
XM_017021539.1 XP_016877028.1 10 1017
XM_017021540.1 XP_016877029.1 9 886
XM_017021541.1 XP_016877030.1 10 1024
XM_017021542.1 XP_016877031.1 8 790
XM_017021543.1 XP_016877032.1 9 894
XM_017021544.1 XP_016877033.1 11 1213
GenBank mRNA AF009746.1 - 12 1274
AK291332.1 - 12 1245
AK297934.1 - 10 1110
AK308382.1 - 11 1128
BC012815.2 - 12 1229 2960427
BT007412.1 - 12 1223
BX247984.1 - 9 949
CR457104.1 - 12 1223

View Full Product Details