Gene Transcripts
- Gene Symbol
- PTPN18
- Entrez Gene ID
- 26469
- Gene Name
- protein tyrosine phosphatase, non-receptor type 18
- Gene Aliases
- BDP1, PTP-HSCF
- Chromosome Location
- Chr. 2: 130356007 - 130375409 on Build GRCh38
- UniGene ID
- Hs.516390
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001142370.1 |
NP_001135842.1
|
3, 4 |
319 |
|
NM_014369.3 |
NP_055184.2
|
7, 8 |
640 |
|
XM_006712416.3 |
XP_006712479.1
|
3, 4 |
290 |
|
XM_006712417.1 |
XP_006712480.1
|
7, 8 |
635 |
|
XM_017003798.1 |
XP_016859287.1
|
4, 5 |
379 |
|
GenBank mRNA |
AK303804.1 |
- |
3, 4 |
242 |
|
BC024280.1 |
- |
3, 4 |
281 |
|
BC041562.1 |
- |
3, 4 |
281 |
|
BC052800.1 |
- |
3, 4 |
281 |
5809095 |
BI261717.1 |
- |
3, 4 |
282 |
|
X79568.1 |
- |
7, 8 |
582 |
|
View Full Product Details