Gene Transcripts

Gene Symbol
AVEN
Entrez Gene ID
57099
Gene Name
apoptosis and caspase activation inhibitor
Gene Aliases
PDCD12
Chromosome Location
Chr. 15: 33858602 - 34074877 on Build GRCh38
UniGene ID
Hs.555966
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020371.2 NP_065104.1 2 445
XM_011521818.2 XP_011520120.1 3 628
XM_011521819.2 XP_011520121.1 2 671
XM_011521820.1 XP_011520122.1 2 146
GenBank mRNA AF283508.1 - 2 441
BC010488.1 - 2 439 2900586
BC063533.1 - 2 429 4425400
BU855198.1 - 519

View Full Product Details