Gene Transcripts

Gene Symbol
Arf2
Entrez Gene ID
11841
Gene Name
ADP-ribosylation factor 2
Gene Aliases
Chromosome Location
Chr. 11: 103966718 - 103985337 on Build GRCm38
UniGene ID
Mm.5061
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK031259.1 - 3 291
AK042977.1 - 3 331
AK053123.1 - 3 749
AK077591.1 - 3 304
AK167133.1 - 3 301
AK167601.1 - 3 305
AK167653.1 - 3 304
AK169094.1 - 3 301
AK169412.1 - 3 301
BC010487.1 - 3 584 3483748
D87899.1 - 3 296

View Full Product Details