Gene Transcripts
- Gene Symbol
- Adra2c
- Entrez Gene ID
- 11553
- Gene Name
- adrenergic receptor, alpha 2c
- Gene Aliases
- Adra-2c, [a]2C, alpha2-C4, alpha2C
- Chromosome Location
- Chr. 5: 35278566 - 35281763 on Build GRCm38
- UniGene ID
- Mm.407152
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007418.3 |
NP_031444.2
|
1 |
3102 |
|
View Full Product Details