Gene Transcripts

Gene Symbol
Atpif1
Entrez Gene ID
11983
Gene Name
ATPase inhibitory factor 1
Gene Aliases
Atpi, IF(1), If1
Chromosome Location
Chr. 4: 132530555 - 132535409 on Build GRCm38
UniGene ID
Mm.2171
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF002718.1 - 3 250
AK008346.1 - 3 248
AK152389.1 - 4 1265
AV462016.1 - 3 311
BC012680.1 - 3 309 4217188
BU055998.1 - 3 237
CB848021.2 - 3 273

View Full Product Details