Gene Transcripts

Gene Symbol
Ncoa2
Entrez Gene ID
17978
Gene Name
nuclear receptor coactivator 2
Gene Aliases
D1Ertd433e, GRIP-1, Grip1, KAT13C, NCoA-2, SRC-2, TIF2, bHLHe75
Chromosome Location
Chr. 1: 13139105 - 13374691 on Build GRCm38
UniGene ID
Mm.2537
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF000582.1 - 13 2947
AK052117.1 - 13 3018
CK637131.1 - 1 51
U39060.1 - 13 2999

View Full Product Details