Gene Transcripts

Gene Symbol
Trim21
Entrez Gene ID
20821
Gene Name
tripartite motif-containing 21
Gene Aliases
Ro52, Ssa1
Chromosome Location
Chr. 7: 102557920 - 102565482 on Build GRCm38
UniGene ID
Mm.321227
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC010580.1 - 8 1101
CT010336.1 - 6 986
L27990.1 - 8 1126

View Full Product Details