Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001171739.1 | NP_001165210.1 | 1 | 372 | |
NM_009736.3 | NP_033866.4 | 1 | 372 | ||
GenBank mRNA | AF019889.1 | - | 1 | 158 | |
AF022223.1 | - | 1 | 313 | ||
AK154469.1 | - | 1 | 322 | ||
AK171094.1 | - | 1 | 322 | ||
BC069918.1 | - | 1 | 309 | 4979705 | |
BC093509.1 | - | 1 | 283 | 4978330 | |
CA321856.1 | - | 1 | 359 | ||
CX225744.1 | - | 1 | 372 |