Gene Transcripts

Gene Symbol
Pam
Entrez Gene ID
18484
Gene Name
peptidylglycine alpha-amidating monooxygenase
Gene Aliases
PHM
Chromosome Location
Chr. 1: 97821091 - 98095632 on Build GRCm38
UniGene ID
Mm.441453
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013626.3 NP_038654.2 6 840
GenBank mRNA AK051649.1 - 6 830
U79523.1 - 6 725

View Full Product Details