Gene Transcripts

Gene Symbol
Vezf1
Entrez Gene ID
22344
Gene Name
vascular endothelial zinc finger 1
Gene Aliases
AI848691, AW046909, db1
Chromosome Location
Chr. 11: 88068256 - 88084729 on Build GRCm38
UniGene ID
Mm.46628
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016686.4 NP_057895.2 2 310
GenBank mRNA AF104410.3 - 2 310

View Full Product Details