Gene Transcripts

Gene Symbol
Bysl
Entrez Gene ID
53414
Gene Name
bystin-like
Gene Aliases
Bys, Enp1
Chromosome Location
Chr. 17: 47599331 - 47611492 on Build GRCm38
UniGene ID
Mm.472101
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016859.3 NP_058555.3 3, 4 896
GenBank mRNA AF007802.1 - 1, 2 125
AK019239.1 - 3, 4 631
AK032993.1 - 3, 4 628
AK049195.1 - 3, 4 889
AK051704.1 - 3, 4 886
AK083033.1 - 3, 4 883
AK143027.1 - 3, 4 897
AK168301.1 - 3, 4 634
BC017530.1 - 3, 4 623 4925307

View Full Product Details