Gene Transcripts
- Gene Symbol
- Styx
- Entrez Gene ID
- 56291
- Gene Name
- serine/threonine/tyrosine interaction protein
- Gene Aliases
- 0610039A20Rik, hStyxb
- Chromosome Location
- Chr. 14: 45351186 - 45376884 on Build GRCm38
- UniGene ID
- Mm.202561
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019637.3 |
NP_062611.2
|
2, 3 |
385 |
|
GenBank mRNA |
AK002822.1 |
- |
|
327 |
|
AK030122.1 |
- |
2, 3 |
385 |
|
AK042741.1 |
- |
2, 3 |
349 |
|
AK046970.1 |
- |
2, 3 |
362 |
|
AK154136.1 |
- |
2, 3 |
480 |
|
AK166043.1 |
- |
2, 3 |
368 |
|
BC038608.1 |
- |
2, 3 |
335 |
5012303 |
BC085089.1 |
- |
2, 3 |
332 |
6492329 |
BC096674.1 |
- |
2, 3 |
393 |
5144003 |
U34973.1 |
- |
2, 3 |
353 |
|
View Full Product Details