Gene Transcripts

Gene Symbol
Ammecr1
Entrez Gene ID
56068
Gene Name
Alport syndrome, mental retardation, midface hypoplasia and elliptocytosis chromosomal region gene 1
Gene Aliases
6230420G18Rik
Chromosome Location
Chr. X: 142853474 - 142966728 on Build GRCm38
UniGene ID
Mm.143724
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019496.3 NP_062369.1 3 691
GenBank mRNA AJ243485.1 - 3 691
AK051736.1 - 3 691

View Full Product Details