Gene Transcripts

Gene Symbol
Astn2
Entrez Gene ID
56079
Gene Name
astrotactin 2
Gene Aliases
1d8, Astnl, bM452J22.1
Chromosome Location
Chr. 4: 65380803 - 66404530 on Build GRCm38
UniGene ID
Mm.445312
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019514.3 NP_062387.2 7, 8 1689
NM_207109.2 NP_996992.1 8, 9 1845
GenBank mRNA AF116573.1 - 5, 6 857
AK041160.1 - 9, 10 1749

View Full Product Details