Gene Transcripts
- Gene Symbol
- Astn2
- Entrez Gene ID
- 56079
- Gene Name
- astrotactin 2
- Gene Aliases
- 1d8, Astnl, bM452J22.1
- Chromosome Location
- Chr. 4: 65380803 - 66404530 on Build GRCm38
- UniGene ID
- Mm.445312
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF116573.1 |
- |
20 |
3813 |
|
View Full Product Details